5-Fluoroindole
Glass bottle for solids, 5 g
Catalog # FLUO006240-5G
Marketsource
Supplier: FLUOROCHEM
Jump to:
Product Details & Documents
Synonyms: 5-Fluoro-1H-indole
- CAS Number:399-52-0
- Formula:C₈H₆FN
- MDL Number:MFCD00005671
- Molecular Weight:135.14 g/mol
- Boiling Pt:85 °C (0,5 mmHg)
- Boiling Pt:85 °C (0.5 mmHg)
- Melting Pt:46 °C
- Storage Temperature:Refrigerator
- EINECS:206-917-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



