3-Fluoropyrocatechol
1 g
Catalog # FLUO006210-1G
Marketsource
Supplier: FLUOROCHEM
Jump to:
Product Details & Documents
Synonyms: 3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
- CAS Number:363-52-0
- Formula:C₆H₅FO₂
- MDL Number:MFCD00042582
- Molecular Weight:128.1 g/mol
- Boiling Pt:218 °C (1013 hPa)
- Melting Pt:71 °C
- Density:1,514 g/cm³ (20 °C)
Specifications
Specifications
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



