N-Hydroxysuccinimide
100 g
Catalog # FLUO005022-100G
Marketsource
Supplier: FLUOROCHEM
N-Hydroxysuccinimide
Catalog # FLUO005022-100G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 6066-82-6
Jump to:
Product Details & Documents
Synonyms: 1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu
- CAS Number:6066-82-6
- Formula:C₄H₅NO₃
- MDL Number:MFCD00005516
- Molecular Weight:115.09 g/mol
- Boiling Pt:363 °C (1013 hPa)
- Melting Pt:93…95 °C
- Storage Temperature:Refrigerator
- EINECS:228-001-3
Specifications
Specifications
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



