3',4'-Dihydroxyacetophenone
Glass bottle for solids, 5 g
Catalog # FLUO002544-5G
Marketsource
Supplier: FLUOROCHEM
3',4'-Dihydroxyacetophenone
Catalog # FLUO002544-5G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 1197-09-7
Jump to:
Product Details & Documents
Synonyms: 1-(3,4-Dihydroxyphenyl)ethan-1-one, Acetopyrocatechol
- CAS Number:1197-09-7
- Formula:C₈H₈O₃
- MDL Number:MFCD00016439
- Molecular Weight:152.15 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



