2-Fluorophenol
Glass bottle for solids, 100 g
Catalog # FLUO001290-100G
Marketsource
Supplier: FLUOROCHEM
Jump to:
Product Details & Documents
Synonyms: o-Fluorophenol
- CAS Number:367-12-4
- Formula:C₆H₅FO
- MDL Number:MFCD00002155
- Molecular Weight:112.1 g/mol
- Boiling Pt:151 °C (1013 hPa)
- Melting Pt:16 °C
- Density:1.22 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:206-681-2
- UN:2810
- ADR:6.1,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



