VISOCOLOR® Colorimetric Test, Nitrate
VISOCOLOR® alpha Nitrate, Reagents for 100 tests and color chart in a blister, 2 • 8 • 15 • 30 • 50 mg/L (NO₃⁻), Azo dye, Visual, 154×93×35 mm (including packaging), Hazardous material: Yes, 100, 1 KIT
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Reagents for the determination of nitrate in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Specifications
Specifications
- Chemical method:Azo dye
- Pk:1 KIT
- Description:VISOCOLOR® alpha Nitrate
- Dimensions:154×93×35 mm (including packaging)
- Type:Hazardous material: Yes
- For unit:Visual
- Tests per kit:100
- Range:2 • 8 • 15 • 30 • 50 mg/L (NO₃⁻)
- Contents:Reagents for 100 tests and color chart in a blister
- Cat. No.:MANA935065
- Platform:Colorimetric test kits
- Model:MANA935065
- Shelf life (months):18
- Storage temperature:15…25 °C
- Weight:71,5 g (including packaging)



