Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Chymostatin

Glass bottle for solids, 5 mg
  BACE4030514.0005
Marketsource
Chymostatin
  BACE4030514.0005
Marketsource
 :  
Chymostatin

Glass bottle for solids, 5 mg

 :  
 
£89.97
   
1 * 5 mg
 
 

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Cat. No.:
    BACE4030514.0005
  • Protein/peptide name:
    Chymostatin
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00071059
  • CAS No.:
    9076-44-2