cis-4-Hydroxy-D(+)-proline
Glass bottle for solids, 25 g
Catalog # BACE4010450.0025
Marketsource
Supplier: BACHEM BIOCHEMICA
cis-4-Hydroxy-D(+)-proline
Catalog # BACE4010450.0025
Supplier: BACHEM BIOCHEMICA
Marketsource
CAS Number: 2584-71-6
Jump to:
Product Details & Documents
Synonyms: (R)-(+)-cis-4-Hydroxyproline, cis-4-Hydroxy-D-proline, H-D-cis-Hyp-OH, H-cis-D-Hyp-OH, D(+)-cis-4-Hydroxyproline
- CAS Number:2584-71-6
- Formula:C₅H₉NO₃
- MDL Number:MFCD00005252
- Molecular Weight:131.13 g/mol
- EINECS:219-963-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



