Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
cis-4-Hydroxy-D(+)-proline

cis-4-Hydroxy-D(+)-proline

Glass bottle for solids, 1 g
Catalog # BACE4010450.0001
Marketsource
Supplier:   BACHEM BIOCHEMICA
cis-4-Hydroxy-D(+)-proline
Catalog # BACE4010450.0001
Supplier:  BACHEM BIOCHEMICA
Marketsource
CAS Number:  2584-71-6
cis-4-Hydroxy-D(+)-proline

Glass bottle for solids, 1 g

CAS Number:  2584-71-6
Price
£62.92
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (R)-(+)-cis-4-Hydroxyproline, cis-4-Hydroxy-D-proline, H-D-cis-Hyp-OH, H-cis-D-Hyp-OH, D(+)-cis-4-Hydroxyproline
  • CAS Number:2584-71-6
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00005252
  • Molecular Weight:131.13 g/mol
  • EINECS:219-963-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.