VISOCOLOR® Colorimetric Test, Nitrate
VISOCOLOR® ECO Nitrate, refill pack, test no.5-41; Measuring range on PF-12Plus. Range on other photometers can be different, Reagents for 110 tests in a box, 0 • 1 • 3 • 5 • 10 • 20 • 30 • 50 • 70 • 90 • 120 mg/L NO₃⁻; 4 - 60 mg/L (NO³⁻), Azo dye, PF-12Plus; PF-3 Soil; Advance; PF-3 Fish; Visual, 170×115×70 mm (including packaging), Hazardous material: Yes, 110, 1 KIT
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Reagents for the determination of nitrate in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Specifications
Specifications
- Chemical method:Azo dye
- Pk:1 KIT
- Description:VISOCOLOR® ECO Nitrate, refill pack, test no.5-41; Measuring range on PF-12Plus. Range on other photometers can be different
- Dimensions:170×115×70 mm (including packaging)
- Type:Hazardous material: Yes
- For unit:PF-12Plus; PF-3 Soil; Advance; PF-3 Fish; Visual
- Tests per kit:110
- Range:0 • 1 • 3 • 5 • 10 • 20 • 30 • 50 • 70 • 90 • 120 mg/L NO₃⁻; 4 - 60 mg/L (NO³⁻)
- Contents:Reagents for 110 tests in a box
- Cat. No.:MANA931241
- Platform:Colorimetric test kits
- Model:MANA931241
- Shelf life (months):18
- Storage temperature:15…25 °C
- Weight:113,5 g (including packaging)



