VISOCOLOR® Colorimetric Test, Nitrate
VISOCOLOR® ECO Nitrate, test no.5-41; Measuring range on PF-12Plus. Range on other photometers can be different, Reagents for 110 tests, color chart and accessories in a box, 0 • 1 • 3 • 5 • 10 • 20 • 30 • 50 • 70 • 90 • 120 mg/L NO₃⁻; 4 - 60 mg/L (NO³⁻), Azo dye, PF-12Plus; PF-3 Soil; Advance; PF-3 Fish; Visual, 168×112×40 mm (including packaging), Hazardous material: Yes, 110, 1 KIT
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Reagents for the determination of nitrate in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Specifications
Specifications
- Chemical method:Azo dye
- Pk:1 KIT
- Description:VISOCOLOR® ECO Nitrate, test no.5-41; Measuring range on PF-12Plus. Range on other photometers can be different
- Dimensions:168×112×40 mm (including packaging)
- Type:Hazardous material: Yes
- For unit:PF-12Plus; PF-3 Soil; Advance; PF-3 Fish; Visual
- Tests per kit:110
- Range:0 • 1 • 3 • 5 • 10 • 20 • 30 • 50 • 70 • 90 • 120 mg/L NO₃⁻; 4 - 60 mg/L (NO³⁻)
- Contents:Reagents for 110 tests, color chart and accessories in a box
- Cat. No.:MANA931041
- Platform:Colorimetric test kits
- Model:MANA931041
- Shelf life (months):18
- Storage temperature:15…25 °C
- Weight:202,74 g (including packaging)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



