Dropper tip caps, WHEATON®
15-415, Blue
Catalog # WHEAW242514-A
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Specifications
- Thread:15-415
- Seq:34
- Cap colour:Blue