Cation exchange chromatography columns, TSK-GEL® SP-5PW
Cation exchange chromatography column, SP-5PW, Analytical, SP-5PW, Glass, 1000 Å, 13 µm, 20,00 mm, 150 mm, Polymer
TOYO0014017
TSK-GEL® SP-5PW are polymer based column packed with porous hydrophilic polymer beads which are surface modified with a strong cation exchanger. Used for the separation and analysis of proteins, peptides, and other biologically active molecules.
- Column Format:Analytical
- Ø int.:20,00 mm
- Particle Size:13 µm
- Pore size:1000 Å
- Description:Cation exchange chromatography column, SP-5PW
- Length:150 mm
- Seq:05
- Column hardware:Glass
- Phase:SP-5PW
- Packing material:Polymer



