Cation exchange chromatography columns, TSK-GEL® SP-5PW
Guardgel kit, Guard column, SP-5PW, Glass, -, 20 µm
TOYO0008807
TSK-GEL® SP-5PW are polymer based column packed with porous hydrophilic polymer beads which are surface modified with a strong cation exchanger. Used for the separation and analysis of proteins, peptides, and other biologically active molecules.
- Column Format:Guard column
- Particle Size:20 µm
- Pore size:-
- Description:Guardgel kit
- Seq:08
- Column hardware:Glass
- Phase:SP-5PW