Cation exchange chromatography columns, TSK-GEL® SP-5PW
Cation exchange chromatography column, SP-5PW, Analytical, SP-5PW, Stainless steel, 1000 Å, 10 µm, 7,50 mm, 75 mm, Polymer
Catalog # TOYO0007161
Supplier: TOSOH Bioscience
Cation exchange chromatography columns, TSK-GEL® SP-5PW
Catalog # TOYO0007161
Supplier: TOSOH Bioscience
Jump to:
Product Details & Documents
TSK-GEL® SP-5PW are polymer based column packed with porous hydrophilic polymer beads which are surface modified with a strong cation exchanger. Used for the separation and analysis of proteins, peptides, and other biologically active molecules.
Specifications
- Column Format:Analytical
- Ø int.:7,50 mm
- Particle Size:10 µm
- Pore size:1000 Å
- Description:Cation exchange chromatography column, SP-5PW
- Length:75 mm
- Seq:03
- Column hardware:Stainless steel
- Phase:SP-5PW
- Packing material:Polymer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



