API hydrometers
API hydrometer, 29 − 41 °API, 335 mm, Soda-lime glass
Catalog # STAN12355/04X
Supplier: STANHOPE SETA
Jump to:
Product Details & Documents
Quality control instruments made of soda lime-silicate glass with a linear coefficient of expansion.
Designed for use in liquids of low surface tension (15 to 34 dynes/cm).
Certifications: ASTM - IP or BS specifications for determining the relative density, specific or API gravity of crude or liquid petroleum products.
Specifications
- Description:API hydrometer
- Length:335 mm
- Material:Soda-lime glass
- Seq:003
- Range:29 − 41 °API