Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Accessories for low temperature viscometer baths, mini
Thermometers, for kinetic viscosity bath, with the range of 98,6 - 101,4 °C, consist of ice point scale and 0,02°C subdivisions for easy reading within 0,01 °C
Catalog # STAN83624-0
Supplier:   STANHOPE SETA
Accessories for low temperature viscometer baths, mini
Catalog # STAN83624-0
Supplier:  STANHOPE SETA
Accessories for low temperature viscometer baths, mini
Accessories for low temperature viscometer baths, mini
Thermometers, for kinetic viscosity bath, with the range of 98,6 - 101,4 °C, consist of ice point scale and 0,02°C subdivisions for easy reading within 0,01 °C
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Thermometers, for kinetic viscosity bath, with the range of 98,6 - 101,4 °C, consist of ice point scale and 0,02°C subdivisions for easy reading within 0,01 °C
  • Seq:
    03