Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Pressure vessels

Pressure vessels

10 l, Dispensing pressure vessel, for holding liquid for filtration
Catalog # MILFXX6700P10
Supplier:   Merck
Pressure vessels
Catalog # MILFXX6700P10
Supplier:  Merck
Pressure vessels
Pressure vessels

10 l, Dispensing pressure vessel, for holding liquid for filtration

Price
£1,830.00
Log in for availability
1 * 1 items
Product Details & Documents

Dispensing pressure vessels hold liquid for filtration through pressure-operated filter holders, if connected to an external pressure source. Used for large-volume filtration, reservoir for buffer or solvent dispensing.

  • Made from stainless steel 316
  • Closures are secured by cam-lock handle
  • Vessels are autoclavable when vented
  • Fluoroelastomer gaskets and O-rings used
  • Molded neoprene rubber base

Max. operating pressure: 6,9 bar

Max. operating temperature: 121 °C

Certifications: Meets ASME®-UM Code requirements.

Specifications
  • Capacity:
    10 l
  • Description:
    Dispensing pressure vessel, for holding liquid for filtration
  • Seq:
    03
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • O-ring, fluoroelastomer A
    O-ring, fluoroelastomer A
    MILFXX6700059
    Log in for availability
    1 * 1 items - £134.00

Product Information

  • Catalog #
    Availability
    Price
  • Bubble point test apparatus
    Bubble point test apparatus
    MILFXX6700L55
    Log in for availability
    1 * 1 items - £1,440.00

Product Information

  • Catalog #
    Availability
    Price
  • Pressure tubing, PVC, I.Ø 12,7 mm
    Pressure tubing, PVC, I.Ø 12,7 mm
    MILFXX6700035
    Log in for availability
    1 * 1 items - £111.00

Product Information

  • Catalog #
    Availability
    Price
  • Pressure vessel accessory kit
    Pressure vessel accessory kit
    MILFXX67000PK
    Log in for availability
    1 * 1 items - £1,550.00

Product Information

  • Catalog #
    Availability
    Price
  • Plug, stainless steel, 6,35 mm NPT
    Plug, stainless steel, 6,35 mm NPT
    MILFYY1301009
    Log in for availability
    1 * 1 items - £47.40

Product Information

  • Catalog #
    Availability
    Price
  • Quick-release nipple and coupling, 6,35 mm NPTM
    Quick-release nipple and coupling, 6,35 mm NPTM
    MILFXX6700030
    Log in for availability
    1 * 1 items - £124.00

Product Information

  • Catalog #
    Availability
    Price
  • Pressure gauge, 6,35 mm ASME
    Pressure gauge, 6,35 mm ASME
    MILFXX6700L15
    Log in for availability
    1 * 1 items - £238.00

Product Information

  • Catalog #
    Availability
    Price
  • Vent/relief valve, 7 bar, ASME
    Vent/relief valve, 7 bar, ASME
    MILFXX6700L24
    Log in for availability
    1 * 1 items - £251.00

Product Information

  • Catalog #
    Availability
    Price
  • Adapter, M, 6,35 mm NPT to 6,35 - 12,7 mm I.Ø
    Adapter, M, 6,35 mm NPT to 6,35 - 12,7 mm I.Ø
    MILFXX6700L11
    Log in for availability
    1 * 2 items - £272.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.