Pressure vessels
5 l, Dispensing pressure vessel, for holding liquid for filtration
Catalog # MILFXX6700P05
Supplier: Merck
Jump to:
Product Details & Documents
Dispensing pressure vessels hold liquid for filtration through pressure-operated filter holders, if connected to an external pressure source. Used for large-volume filtration, reservoir for buffer or solvent dispensing.
- Made from stainless steel 316
- Closures are secured by cam-lock handle
- Vessels are autoclavable when vented
- Fluoroelastomer gaskets and O-rings used
- Molded neoprene rubber base
Max. operating pressure: 6,9 bar
Max. operating temperature: 121 °C
Certifications: Meets ASME®-UM Code requirements.
Specifications
- Capacity:5 l
- Description:Dispensing pressure vessel, for holding liquid for filtration
- Seq:02
Accessories
Product Information
- Catalog #AvailabilityPrice
- O-ring, fluoroelastomer A
Product Information
- Catalog #AvailabilityPrice
- Bubble point test apparatus
Product Information
- Catalog #AvailabilityPrice
- Pressure tubing, PVC, I.Ø 12,7 mm
Product Information
- Catalog #AvailabilityPrice
- Pressure vessel accessory kit
Product Information
- Catalog #AvailabilityPrice
- Plug, stainless steel, 6,35 mm NPT
Product Information
- Catalog #AvailabilityPrice
- Quick-release nipple and coupling, 6,35 mm NPTMQuick-release nipple and coupling, 6,35 mm NPTMMILFXX6700030Log in for availability1 * 1 items - £124.00
Product Information
- Catalog #AvailabilityPrice
- Pressure gauge, 6,35 mm ASME
Product Information
- Catalog #AvailabilityPrice
- Vent/relief valve, 7 bar, ASME
Product Information
- Catalog #AvailabilityPrice
- Adapter, M, 6,35 mm NPT to 6,35 - 12,7 mm I.ØAdapter, M, 6,35 mm NPT to 6,35 - 12,7 mm I.ØMILFXX6700L11Log in for availability1 * 2 items - £272.00



