Dropping funnels, conical shape, without pressure balance
500 ml, 29/32 NS, 29/32 NS
Catalog # 201-0912
Supplier: Lenz Laborglas
Dropping funnels, conical shape, without pressure balance
Catalog # 201-0912
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
Borosilicate glass 3.3.
- Screw thread retaining device
- With inscription label, without graduation
- Minimal thermal expansion giving relatively high resistance to temperature changes
With NS socket, NS cone and stopcock with glass or PTFE plug.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Cone size:29/32 NS
- Socket size:29/32 NS
- Capacity:500 ml