Distilling receivers, Bernauer
19/26 NS
Catalog # LENZ05288002
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling receivers acc. to Bernauer (modified), with three cones NS 14/23, vacuum connection with glass olive. With flasks 100 ml capacity or without flasks available.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Socket size:19/26 NS