Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
UV blocking eyewear
UV blocking eyewear
Goggles (anti-UV, anti-fog)
Catalog # 730-1034
UV blocking eyewear
Catalog # 730-1034
UV blocking eyewear
UV blocking eyewear
Goggles (anti-UV, anti-fog)
Price
£42.50
Log in for availability
1 * 1 items
Product Details & Documents
Eye and face protection for applications involving short wave ultraviolet sources. Available in three styles.
  • Spectacles are made of impact resistant PC; the top and side-shield design provides full UV protection
  • Goggles are constructed of specially formulated plastic and feature a flexible rubber trim which fits comfortably on the face and over most prescription spectacles, allowing indirect ventilation
ANSI-Z87.1 1986
Certifications: ANSI-Z87.1 1986
Documents
Specifications
  • Description:
    Goggles (anti-UV, anti-fog)
  • Seq:
    02