Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Evaporating flasks
Evaporating flasks
100 ml, - NS, EL-flange, uncoated
Catalog # 531-1971
Supplier:   BUCHI
Evaporating flasks
Catalog # 531-1971
Supplier:  BUCHI
Evaporating flasks
Evaporating flasks
100 ml, - NS, EL-flange, uncoated
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® rotary evaporators. Centre neck. Pear-shaped with ground joint socket or EL-flange.
Specifications
  • Socket size:
    - NS
  • Type:
    EL-flange, uncoated
  • Capacity:
    100 ml
  • Cat. No.:
    531-1971