Biopsy processing cassettes, Histosette® II
Yellow
Catalog # 631-9659
Supplier: DELTALAB
Jump to:
Product Details & Documents
Made of acetal polymer.1 mm square openings to maximise fluid exchange and to ensure proper drainage.
- With 45° angle writing area
- Suitable for automatic labellers
- No need to break the lid and then put it in place, as it is supplied pre-mounted
- Can be easily opened and closed with just one hand
Specifications
- Colour:Yellow
- Seq:03