Order Entry
United Kingdom
ContactUsLinkComponent
Tray 350x250x40mm gsp,bowls - abswhite. in impact-resistant, stable design, with smooth surface. temperature resistant up to +85 ° c , mass 350x250x 40mm 1 * 1 items
Catalog # 216-1250
Supplier:  SEMADENI
Tray 350x250x40mm gsp,bowls - abswhite. in impact-resistant, stable design, with smooth surface. temperature resistant up to +85 ° c , mass 350x250x 40mm 1 * 1 items
Catalog # 216-1250
Supplier:  SEMADENI
Frequently Bought Together