Iron(III) phosphate hydrate
Plastic bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
Iron(III) phosphate hydrate
Catalog # 31106.30
Supplier: Thermo Fisher Scientific
CAS Number: 51833-68-2
Jump to:
Product Details & Documents
CAS No.: 51833-68-2 Molecular Formula: FePO4.xH2O Formula Weight: 150.82(anhy) Physical Form: Powder Appearance: Off-white
Synonyms: Iron phosphate hydrate, Iron (III) orthophosphate hydrate
- CAS Number:51833-68-2
- Formula:FeO₄P·xH₂O
- MDL Number:MFCD00016096
- Molecular Weight:150.82 g/mol (anhy)
- Storage Temperature:Ambient
- EINECS:233-149-7
Documents
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



