Test kits, chlorine, Spectroquant®, Supelco®
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*4 Reagents for free chlorine testing are reagent Cl₂-1 1.00086.0001 and reagent Cl₂-2 1.00087.0001 together with empty cell and accessories 1.00089.0001
Reagents for total chlorine are reagent Cl₂-1 1.00086.0001, reagent Cl₂-2 1.00087.0001 and reagent Cl₂-3 1.00088.0001 together with empty cell and accessories 1.00089.0001
"*7 This method is officially recognised by the USEPA as an alternative method for the investigation of the following:
*7-1 wastewater
*7-2 drinking water
*7-3 respectively drinking water and waste water
Documents
Specifications
Specifications
- Chemical method:DPD
- Pk:200 Tests
- Description:Chlorine test (free chlorine)*<sup>7-2</sup>
- For unit:Prove, Nova 60, Multy, Move
- Range:0,010 - 6,00 mg/L Cl₂
- Cat. No.:1.00598.0002
- M-Clarity quality level:MQ100
Specifications
Accessories
Product Information
- Catalog #For unitPkAvailabilityPrice
- Chlorine, cells and accessories for photometric Cl₂ measurement*4Chlorine, cells and accessories for photometric Cl₂ measurement*41.00089.0001Prove, Nova 6025Log in for availability1 * 25 items - £83.40