4'-Hydroxyacetophenone for synthesis, Sigma-Aldrich®
Plastic bottle, 250 g
Supplier: Merck
4'-Hydroxyacetophenone for synthesis, Sigma-Aldrich®
Catalog # 8.04311.0250
Supplier: Merck
CAS Number: 99-93-4
Jump to:
Product Details & Documents
- CAS Number:99-93-4
- Formula:C₈H₈O₂
- MDL Number:MFCD00002359
- Molecular Weight:136.15 g/mol
- Boiling Pt:313 °C (1013 hPa)
- Melting Pt:105…108 °C
- Density:1.139 g/cm³ (25 °C)
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:202-802-8
Documents
Specifications
Specification Test Results
- Solubility:9.9 g/l (22 °C)
- Melting Point:105 - 108 °C
- Molar Mass:136.15 g/mol
- Flash Point:166 °C
- Synonyms:4-Acetylphenol
- Hill Formula:C8H8O2
- Chemical Formula:4-(OH)C6H4COCH3
Specifications
- Pack type:Plastic bottle
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



