Origami B Competent Cells 1 * 0,4 mL
Catalog # 70836-3
Supplier: Merck Millipore (Novagen)
Frequently Bought Together
Deep well storage plates, 96-well, 2,2 ml
Supplier: THERMO MATRIX TECHNOLOGIES
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
MANTIS CF CAP - FALCON 50ML CONICAL TUBE, SILICONE HARNESS 1 * 2 items
Supplier: MARKET SOURCE PART PROCESS
Scoops for foodstuffs, SteriPlast®
Supplier: BURKLE
Containers, wide mouth, with screw caps
Supplier: KARTELL
Whatman™ Protran® BA85 & BA83 NC Western Blotting Membranes, Nitrocellulose
Supplier: Whatman products (Cytiva)
Cryoboxes, for microtubes, B30/B32-series
Supplier: GLW
Cryoboxes, 133×133 mm
Supplier: RATIOLAB
Tubes Cryo.S™ 4 and 5 ml
Supplier: Greiner Bio-One
Cap sealing mats for VersiPlate 96-well PCR strip tube plates, VersiCap™ Mats
Supplier: Thermo Fisher Scientific
Restriction enzyme Nb.Mva1269I 1000 units (10u/µl) 1 * 1 KIT
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Half Size, 136×66,5 mm
Supplier: VWR Collection
Cryoboxes, coloured, Nalgene®
Supplier: Thermo Fisher Scientific
FalconRegistered Mark Round-Bottom Tubes, 14 mL 1 * 500 items
Supplier: STEMCELL Technologies