Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Visual colour kits, total hardness, MQuant®, Supelco®

Visual colour kits, total hardness, MQuant®, Supelco®

Total hardness test, 0,18 - 10 °f, (1 - 100 mg/l CaCO₃), Titriplex® III, MTC, 300 Tests
Catalog # 1.08047.0001
Supplier:   Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Catalog # 1.08047.0001
Supplier:  Merck
CAS Number:  
Visual colour kits, total hardness, MQuant®, Supelco®
Visual colour kits, total hardness, MQuant®, Supelco®

Total hardness test, 0,18 - 10 °f, (1 - 100 mg/l CaCO₃), Titriplex® III, MTC, 300 Tests

CAS Number:  
Price
£143.00
Log in for availability
1 * 300 Tests
Product Details & Documents

Quick tests, visual colour comparison.

  • Results in approximately 5 minutes
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available

MQuant® titrimetric and colorimetric methods:

This range comprises both titrimetric and colorimetric tests for medium concentrations. With its broad range of available test parameters, these quick tests are the method of choice for aquaria, fish farming, aquatic ecology, environmental, cooling and boiler water analysis.

RP= Refill pack

** 1 drop

Delivery information: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.

Specifications

Specifications

  • Chemical method:
    Titriplex® III
  • Pk:
    300 Tests
  • Description:
    Total hardness test
  • Type:
    MTC
  • Range:
    0,18 - 10 °f, (1 - 100 mg/l CaCO₃)
  • Cat. No.:
    1.08047.0001
  • M-Clarity quality level:
    MQ100

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.