Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Embedding and mounting media
Embedding and mounting media
Rapid for permanent specimens, 60-100 mPas, preparative capacity of 2500 slides, 500 ml
Catalog # 1.07960.0500
Supplier:   Merck
Embedding and mounting media
Catalog # 1.07960.0500
Supplier:  Merck
CAS Number:  
Embedding and mounting media
Embedding and mounting media
Rapid for permanent specimens, 60-100 mPas, preparative capacity of 2500 slides, 500 ml
CAS Number:  
Price
£126.00
Log in for availability
1 * 500 mL
Product Details & Documents
The defined hardness and uniformity of the material to be cut are vital requirements in creating consistent quality in cutting. These requirements can be met using a range of suitable substances, such as paraffins, resins, gelatine and celloidin.
Specifications
Specifications
  • Pk:
    500 ml
  • Description:
    Rapid for permanent specimens, 60-100 mPas, preparative capacity of 2500 slides
Specifications