Order Entry
United Kingdom
ContactUsLinkComponent
SGE SYRINGE 500µL REMOVABLE NEEDLE NEEDLE IS STAINLESS STEEL 51 MM LONG 22 GAUGE (0.71 MM OD) WITH LC POINT STYLE AND FITTED WITH A REPEATING ADAPTER
Catalog # 549-0471
SGE SYRINGE 500µL REMOVABLE NEEDLE NEEDLE IS STAINLESS STEEL 51 MM LONG 22 GAUGE (0.71 MM OD) WITH LC POINT STYLE AND FITTED WITH A REPEATING ADAPTER
Catalog # 549-0471
Frequently Bought Together