Order Entry
United Kingdom
ContactUsLinkComponent
SGE SYRINGE 5µL REMOVABLE NEEDLE FOR AGILENT 7673/7683 AUTOSAMPLER. NEEDLE IS STAINLESS STEEL 0.63 MM OD (23 GAUGE) 42 MM LONG WITH A CONE POINT STYLE
Catalog # 562-1017
SGE SYRINGE 5µL REMOVABLE NEEDLE FOR AGILENT 7673/7683 AUTOSAMPLER. NEEDLE IS STAINLESS STEEL 0.63 MM OD (23 GAUGE) 42 MM LONG WITH A CONE POINT STYLE
Catalog # 562-1017
Frequently Bought Together