
Sodium hydroxide ∼32% in aqueous solution, EMPLURA®, Supelco®
1.05587.9200
: Merck
:
- Pack type:Plastic barrel
- Pk:200 L
- Seq:5549
- Solubility:(20 °C) soluble
- Melting Point:9 °C
- Boiling Point:119 °C (1013 hPa)
- Vapor Pressure:8 hPa (20 °C)
- Density:1.35 g/cm3 (20 °C)
- pH:14 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
:
:40 g/mol
:119…135 °C (1013 hPa)
:9…10 °C
:1,35 g/cm³ (20 °C)
:Ambient
:215-185-5
:1824
:8,II
:M-Clarity™ quality level = MQ200
:GHS05
Frequently Bought Together