Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Dextran, M.W. ~ 60,000

Dextran, M.W. ~ 60,000

100 g
Catalog # SERA18692.01
Dextran, M.W. ~ 60,000
Catalog # SERA18692.01
Supplier:  
CAS Number:  9004-54-0
Dextran, M.W. ~ 60,000

100 g

CAS Number:  9004-54-0
Discontinued
Product Details & Documents
  • CAS Number:9004-54-0
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00130935
  • EINECS:232-677-5
Specifications

Specifications

  • Pk:
    100 g

Specifications