Thymidine (2'-deoxythymidine)
Glass bottle for solids, 1 g
Catalog # SERA18600.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
- CAS Number:50-89-5
- Formula:C₁₀H₁₄N₂O₅
- MDL Number:MFCD00006537
- Molecular Weight:242.23 g/mol
- Storage Temperature:Ambient
- EINECS:200-070-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



