Bacitracin
Glass bottle for solids, 2,5 g
Catalog # SERA14420.01
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
- CAS Number:1405-87-4
- Formula:C₆₆H₁₀₃N₁₇O₁₆S₁
- MDL Number:MFCD00062640
- Molecular Weight:1421.60 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:2,5 g
- Cat. No.:SERA14420.01
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



