Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Agar

Agar

Glass bottle for solids, 1 kg
Catalog # SERA11393.04
Marketsource
Supplier:   Serva
Agar
Catalog # SERA11393.04
Supplier:  Serva
Marketsource
CAS Number:  9002-18-0
Agar
Agar

Glass bottle for solids, 1 kg

CAS Number:  9002-18-0
Price
£327.33
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications