Burette clamps
Two burettes
Catalog # 241-2021
Supplier: KARTELL
Jump to:
Product Details & Documents
PP, white, robust. For fast and safe clamping of burettes.
- Clamps fit rods 8 to 14 mm diameter
- Clamps have rubber inserts and stainless steel springs
- Burette scale not obstructed
Specifications
- For:Two burettes