Indicator strips, non bleeding
Indicator strips, non bleeding, 0 - 14
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Non bleeding special indicator dyes that are covalently bound onto the reagent paper and then adhered to plastic strips.
Documents
Certificates
Search by Lot Numberspec_VWRC315052J spec_VWRC315052J spec_VWRC315052J spec_VWRC315082P
Product specification
Specifications
- pH range:0 - 14
- Description:Indicator strips, non bleeding