Mercury(II) bromide 99+%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Mercury(II) bromide 99+%
Catalog # ACRO190495000
Supplier: Thermo Fisher Scientific
CAS Number: 7789-47-1
Jump to:
Product Details & Documents
Synonyms: Mercuric bromide, Mercury dibromide
- CAS Number:7789-47-1
- Formula:Br₂Hg
- MDL Number:MFCD00011040
- Molecular Weight:360.4 g/mol
- Merck Index:13,05900
- Boiling Pt:320 °C (1013 hPa)
- Melting Pt:236 °C
- Density:6,1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:232-169-3
- UN:1634
- ADR:6.1,II
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Assay:≥99.0 %
- Chloride (Cl):≤2000 ppm
- Iron (Fe):≤50 ppm
- Insoluble matter:≤0.05 % (in methanol)
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



