2-Chlorophenol 98+%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1-Chloro-2-hydroxybenzene
- CAS Number:95-57-8
- Formula:C₆H₅ClO
- MDL Number:MFCD00002159
- Molecular Weight:128.56 g/mol
- Merck Index:12,02173
- Boiling Pt:173…175 °C (760 mmHg)
- Melting Pt:8 °C
- Density:1.262 g/cm³
- Flash Pt:63 °C
- Storage Temperature:Ambient
- EINECS:202-433-2
- UN:2021
- ADR:6.1,III
Specifications
Specification Test Results
- Appearance:Clear light yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5580 to 1.5600 (20°C, 589 nm)
- GC:≥98.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



