2-Chlorophenol 98+%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1-Chloro-2-hydroxybenzene
- CAS Number:95-57-8
- Formula:C₆H₅ClO
- MDL Number:MFCD00002159
- Molecular Weight:128.56 g/mol
- Merck Index:12,02173
- Boiling Pt:173…175 °C (760 mmHg)
- Melting Pt:8 °C
- Density:1.262 g/cm³
- Flash Pt:63 °C
- Storage Temperature:Ambient
- EINECS:202-433-2
- UN:2021
- ADR:6.1,III
Specifications
Specification Test Results
- Appearance:Clear light yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5580 to 1.5600 (20°C, 589 nm)
- GC:≥98.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



