Phenyl benzoate 99%
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Benzoic acid phenyl ester
- CAS Number:93-99-2
- Formula:C₁₃H₁₀O₂
- MDL Number:MFCD00003072
- Molecular Weight:198.22 g/mol
- Merck Index:12,07429
- Boiling Pt:299 °C (1013 hPa)
- Melting Pt:68…70 °C
- Density:1,041 g/cm³ (131 °C)
- Storage Temperature:Ambient
- EINECS:202-293-2
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:68 to 70 °C
- GC:≥98.5 %
- Acid value:≤1 mg KOH/g
Specifications
- Pack type:Plastic bottle
- Pk:500 g



