Isovaleric acid 99%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 3-Methylbutyric acid, Isopropyl acetic acid, 3-Methylbutanoic acid
- CAS Number:503-74-2
- Formula:C₅H₁₀O₂
- MDL Number:MFCD00002726
- Molecular Weight:102.13 g/mol
- Merck Index:12,05251
- Boiling Pt:176 °C (760 mmHg)
- Melting Pt:–35 °C
- Density:0.926 g/cm³
- Flash Pt:70 °C
- Storage Temperature:Ambient
- EINECS:207-975-3
- UN:3265
- ADR:8,II
Documents
ACRO15669
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3990 to 1.4070 (20°C, 589 nm)
- GC:≥98.5 %
- Specific gravity:(25°C) 0.921 to 0.93
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



