4'-Hydroxyacetophenone 98%
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
4'-Hydroxyacetophenone 98%
Catalog # ACRO150401000
Supplier: Thermo Fisher Scientific
CAS Number: 99-93-4
Jump to:
Product Details & Documents
- CAS Number:99-93-4
- Formula:C₈H₈O₂
- MDL Number:MFCD00002359
- Molecular Weight:136.15 g/mol
- Boiling Pt:147…148 °C (3 mmHg)
- Melting Pt:107…111 °C
- Density:1,139 g/cm³ (25 °C)
- Flash Pt:166 °C
- Storage Temperature:Ambient
- EINECS:202-802-8
Specifications
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:107 to 111 °C
- GC:≥97.5 %
Specifications
- Pack type:Plastic bottle
- Pk:100 g



