4'-Hydroxyacetophenone 98%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
4'-Hydroxyacetophenone 98%
Catalog # ACRO150400050
Supplier: Thermo Fisher Scientific
CAS Number: 99-93-4
Jump to:
Product Details & Documents
- CAS Number:99-93-4
- Formula:C₈H₈O₂
- MDL Number:MFCD00002359
- Molecular Weight:136.15 g/mol
- Boiling Pt:147…148 °C (3 mmHg)
- Melting Pt:107…111 °C
- Density:1,139 g/cm³ (25 °C)
- Flash Pt:166 °C
- Storage Temperature:Ambient
- EINECS:202-802-8
Documents
Specifications
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:107 to 111 °C
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



