Cyclohexanecarboxylic acid 98%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Cyclohexanecarboxylic acid 98%
Catalog # ACRO146565000
Supplier: Thermo Fisher Scientific
CAS Number: 98-89-5
Jump to:
Product Details & Documents
Synonyms: Hexahydrobenzoic acid
- CAS Number:98-89-5
- Formula:C₇H₁₂O₂
- MDL Number:MFCD00001461
- Molecular Weight:128.17 g/mol
- Boiling Pt:232 °C (1013 hPa)
- Melting Pt:30…32 °C
- Density:1.03 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:202-711-3
Documents
ACRO14656
SDS
Specifications
Specification Test Results
- Appearance:Colorless to white Fused crystalline solid or viscous liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Titration with NaOH:97.5 to 102.5 %
- Elemental Analysis:33.33 to 35.03 % C
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



