Cyclohexanecarboxylic acid 98%
Glass bottle for solids, 1 kg
Supplier: Thermo Fisher Scientific
Cyclohexanecarboxylic acid 98%
Catalog # ACRO146560010
Supplier: Thermo Fisher Scientific
CAS Number: 98-89-5
Jump to:
Product Details & Documents
Synonyms: Hexahydrobenzoic acid
- CAS Number:98-89-5
- Formula:C₇H₁₂O₂
- MDL Number:MFCD00001461
- Molecular Weight:128.17 g/mol
- Boiling Pt:232 °C (1013 hPa)
- Melting Pt:30…32 °C
- Density:1.03 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:202-711-3
Specifications
Specification Test Results
- Appearance:Colorless to white Fused crystalline solid or viscous liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Titration with NaOH:97.5 to 102.5 %
- Elemental Analysis:33.33 to 35.03 % C
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



