3,3'-Thiodipropionic acid 99%
Plastic bottle, 25 g
Supplier: Thermo Fisher Scientific
3,3'-Thiodipropionic acid 99%
Catalog # ACRO138750250
Supplier: Thermo Fisher Scientific
CAS Number: 111-17-1
Jump to:
Product Details & Documents
Synonyms: 3,3-Thiodipropionic acid
- CAS Number:111-17-1
- Formula:C₆H₁₀O₄S
- MDL Number:MFCD00002781
- Molecular Weight:178.21 g/mol
- Merck Index:12,09468
- Boiling Pt:347 °C (1013 hPa)
- Melting Pt:130…133 °C
- Density:1,496 g/cm³ (20 °C)
- Flash Pt:128 °C
- Storage Temperature:Ambient
- EINECS:203-841-3
Documents
ACRO13875
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:130 to 133 °C
- Titration with NaOH:98.5 to 101.5 %
- NMR (1H-NMR):Consistent with structure
Specifications
- Pack type:Plastic bottle
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



